Methyl-6-(trifluoromethyl)-2,3-dihydro-1H-indol-5-yl ether ≥95%
Supplier: Matrix Scientific
047164-500MG
047164-1G
101878-738EA
921.54
USD
101878-738
101878-740
Methyl-6-(trifluoromethyl)-2,3-dihydro-1H-indol-5-yl ether ≥95%
Methyl-6-(trifluoromethyl)-2,3-dihydro-1H-indol-5-yl ether
Hazard: Irritant
Formula:
C₁₀H₁₀F₃NO Melting Pt: 83…84 °C |
MDL Number:
MFCD12498700 CAS Number: 178896-79-2 |
Specification Test Results
Melting point | 83-84°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...