2-Chloro-4,6-diiodo-3-pyridinol ≥95%

Supplier: Matrix Scientific

047160-500MG 047160-1G 047160-5G
101878-720EA 375.38 USD
101878-720 101878-722 101878-724
2-Chloro-4,6-diiodo-3-pyridinol ≥95%
2-Chloro-4,6-diiodo-3-pyridinol

Hazard: Irritant

Formula: C₅H₂ClI₂NO
Melting Pt: 143…145 °C
MDL Number: MFCD12498699
CAS Number: 1171918-88-9

Order Now

Specification Test Results

Melting point 143-145°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR