5-(Benzyloxy)-4-chloro-2-(chloromethyl)pyridine ≥95%
Supplier: Matrix Scientific
047159-500MG
047159-1G
101878-716EA
443.08
USD
101878-716
101878-718
5-(Benzyloxy)-4-chloro-2-(chloromethyl)pyridine ≥95%
5-(Benzyloxy)-4-chloro-2-(chloromethyl)pyridine
Hazard: Irritant
Formula:
C₁₃H₁₁Cl₂NO Melting Pt: 82…84 °C |
MDL Number:
MFCD12498698 CAS Number: 62811-98-7 |
Specification Test Results
Melting point | 82-84°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...