Ethyl-4-hydroxy-2-(2-pyridinyl)-5-pyrimidinecarboxylate ≥95%
Supplier: Matrix Scientific
047156-500MG
047156-1G
047156-5G
101878-700EA
269.23
USD
101878-700
101878-702
101878-704
Ethyl-4-hydroxy-2-(2-pyridinyl)-5-pyrimidinecarboxylate ≥95%
Ethyl-4-hydroxy-2-(2-pyridinyl)-5-pyrimidinecarboxylate
Hazard: Irritant
Formula:
C₁₂H₁₁N₃O₃ MW: 245.24 g/mol Melting Pt: 106…107 °C |
MDL Number:
MFCD00102681 CAS Number: 60060-10-8 |
Specification Test Results
Melting point | 106-107°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...