5-Bromo-2-methylbenzoxazole ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-methyl-1,3-benzoxazole

047153-500MG 047153-1G
101878-692EA 87.69 USD
101878-692 101878-694
5-Bromo-2-methylbenzoxazole ≥95%
5-Bromo-2-methylbenzoxazole

Hazard: Irritant

Formula: C₈H₆BrNO
MW: 212.0400 g/mol
Melting Pt: 66-68°C
MDL Number: MFCD08702771
CAS Number: 5676-56-2

Order Now

Specification Test Results

Melting point 66-68°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR