5-Bromo-2-ethylbenzoxazole ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-ethylbenzo[d]oxazole

047152-500MG 047152-1G 047152-5G
101878-686EA 278.46 USD
101878-686 101878-688 101878-690
5-Bromo-2-ethylbenzoxazole ≥95%
5-Bromo-2-ethylbenzoxazole

Hazard: Irritant

Formula: C₉H₈BrNO
MW: 226.0700 g/mol
Melting Pt: 64-66°C
MDL Number: MFCD09055180
CAS Number: 938458-80-1

Order Now

Specification Test Results

Melting point 64-66°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR