Ethyl-4-hydroxy-2-(trifluoromethyl)-5-pyrimidinecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 4-hydroxy-2-(trifluoromethyl)pyrimidine-5-carboxylate
047151-500MG
047151-1G
047151-5G
101878-680EA
112.31
USD
101878-680
101878-682
101878-684
Ethyl-4-hydroxy-2-(trifluoromethyl)-5-pyrimidinecarboxylate ≥95%
Ethyl-4-hydroxy-2-(trifluoromethyl)-5-pyrimidinecarboxylate
Hazard: Irritant
Formula:
C₈H₇F₃N₂O₃ MW: 236.15 g/mol Melting Pt: 315…316 °C |
MDL Number:
MFCD01571344 CAS Number: 343-67-9 |
Specification Test Results
Melting point | 315-316°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...