2H-1,4-Benzothiazine-2,3(4H)-dione-3-oxime ≥95%
Supplier: Matrix Scientific
047149-500MG
047149-1G
047149-5G
101878-670EA
181.54
USD
101878-670
101878-672
101878-674
2H-1,4-Benzothiazine-2,3(4H)-dione-3-oxime ≥95%
2H-1,4-Benzothiazine-2,3(4H)-dione-3-oxime
Hazard: Irritant
Formula:
C₈H₆N₂O₂S MW: 194.21 g/mol Melting Pt: 270…272 °C |
MDL Number:
MFCD08689794 CAS Number: 903891-96-3 |
Specification Test Results
Melting point | 270-272°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...