6-Chloro-2H-1,4-benzoxazine-2,3(4H)-dione-3-oxime ≥95%
Supplier: Matrix Scientific
047148-500MG
047148-1G
047148-5G
101878-664EA
181.54
USD
101878-664
101878-666
101878-668
6-Chloro-2H-1,4-benzoxazine-2,3(4H)-dione-3-oxime ≥95%
6-Chloro-2H-1,4-benzoxazine-2,3(4H)-dione-3-oxime
Hazard: Irritant
Formula:
C₈H₅ClN₂O₃ MW: 212.59 g/mol Melting Pt: 231…233 °C |
MDL Number:
MFCD08689793 CAS Number: 937602-19-2 |
Specification Test Results
Melting point | 231-233°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...