N-Methyl-5-(trifluoromethyl)-2-pyridinamine ≥95%

Supplier: Matrix Scientific

Synonyms: N-Methyl-5-(trifluoromethyl)pyridin-2-amine, 2-(Methylamino)-5-(trifluoromethyl)pyridine

047145-1G 047145-5G
101878-648EA 136.92 USD
101878-648 101878-650
N-Methyl-5-(trifluoromethyl)-2-pyridinamine ≥95%
N-Methyl-5-(trifluoromethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₇H₇F₃N₂
MW: 176.14 g/mol
Melting Pt: 91…93 °C
MDL Number: MFCD08689790
CAS Number: 937602-15-8

Order Now

Specification Test Results

Melting point 91-93°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR