2H-1,4-Benzoxazine-2,3(4H)-dione-3-oxime ≥95%

Supplier: Matrix Scientific

047144-500MG 047144-1G 047144-5G
101878-642EA 181.54 USD
101878-642 101878-644 101878-646
2H-1,4-Benzoxazine-2,3(4H)-dione-3-oxime ≥95%
2H-1,4-Benzoxazine-2,3(4H)-dione-3-oxime

Hazard: Irritant

Formula: C₈H₆N₂O₃
MW: 178.15 g/mol
Melting Pt: 270…274 °C
MDL Number: MFCD08689789
CAS Number: 903891-95-2

Order Now

Specification Test Results

Melting point 270-274°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR