Ethyl-1,6-dimethyl-2-oxo-4-(3-pyridinyl)-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%

Supplier: Matrix Scientific

047143-500MG 047143-1G 047143-5G
101878-636EA 181.54 USD
101878-636 101878-638 101878-640
Ethyl-1,6-dimethyl-2-oxo-4-(3-pyridinyl)-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Ethyl-1,6-dimethyl-2-oxo-4-(3-pyridinyl)-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₇N₃O₃
MW: 275.31 g/mol
Melting Pt: 124…126 °C
MDL Number: MFCD02972683
CAS Number: 512840-33-4

Order Now

Specification Test Results

Melting point 124-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR