Ethyl-4-(2-chlorophenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%

Supplier: Matrix Scientific

047140-500MG 047140-1G 047140-5G
101878-618EA 181.54 USD
101878-618 101878-620 101878-622
Ethyl-4-(2-chlorophenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Ethyl-4-(2-chlorophenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₇ClN₂O₃
Melting Pt: 142…144 °C
MDL Number: MFCD00205390
CAS Number: 253328-48-2

Order Now

Specification Test Results

Melting point 142-144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR