Ethyl-4-(3-fluorophenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%

Supplier: Matrix Scientific

047135-500MG 047135-1G 047135-5G
101878-588EA 181.54 USD
101878-588 101878-590 101878-592
Ethyl-4-(3-fluorophenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Ethyl-4-(3-fluorophenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₇FN₂O₃
Melting Pt: 169…171 °C
MDL Number: MFCD02324169
CAS Number: 342596-78-5

Order Now

Specification Test Results

Melting point 169-171°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR