Ethyl-4-(4-chlorophenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%

Supplier: Matrix Scientific

047134-500MG 047134-1G 047134-5G
101878-582EA 181.54 USD
101878-582 101878-584 101878-586
Ethyl-4-(4-chlorophenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Ethyl-4-(4-chlorophenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₇ClN₂O₃
Melting Pt: 124…126 °C
MDL Number: MFCD00718037
CAS Number: 302821-62-1

Order Now

Specification Test Results

Melting point 124-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR