Methyl-2-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-methyl-3-oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-6-carboxylate

047130-500MG 047130-1G
101878-566EA 375.38 USD
101878-566 101878-568
Methyl-2-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate ≥95%
Methyl-2-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₁NO₄
MW: 221.21 g/mol
Melting Pt: 159…161 °C
MDL Number: MFCD11100232
CAS Number: 179950-77-7

Order Now

Specification Test Results

Melting point 159-161°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR