5-Bromo-2-(chloromethyl)benzoxazole ≥95%

Supplier: Matrix Scientific

047129-500MG 047129-1G
101878-562EA 461.54 USD
101878-562 101878-564
5-Bromo-2-(chloromethyl)benzoxazole ≥95%
5-Bromo-2-(chloromethyl)benzoxazole

Hazard: Irritant

Formula: C₈H₅BrClNO
Melting Pt: 68-70°C
MDL Number: MFCD10696614
CAS Number: 110704-48-8

Order Now

Specification Test Results

Melting point 68-70°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR