5-Chloro-2-hydroxynicotinonitrile ≥95%
Supplier: Matrix Scientific
047128-500MG
047128-1G
101878-558EA
109.23
USD
101878-558
101878-560
5-Chloro-2-hydroxynicotinonitrile ≥95%
5-Chloro-2-hydroxynicotinonitrile
Hazard: Irritant
Formula:
C₆H₃ClN₂O Melting Pt: 199…201 °C |
MDL Number:
MFCD11100231 CAS Number: 1048913-62-7 |
Specification Test Results
Melting point | 199-201°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...