6-Bromo-2-methyl-2H-1,4-benzoxazin-3(4H)-one ≥95%
Supplier: Matrix Scientific
047127-500MG
047127-1G
101878-554EA
400
USD
101878-554
101878-556
6-Bromo-2-methyl-2H-1,4-benzoxazin-3(4H)-one ≥95%
6-Bromo-2-methyl-2H-1,4-benzoxazin-3(4H)-one
Hazard: Irritant
Formula:
C₉H₈BrNO₂ Melting Pt: 188…190 °C |
MDL Number:
MFCD08446951 CAS Number: 221311-16-6 |
Specification Test Results
Melting point | 188-190°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...