2-Amino-5-bromopyrimidin-4(3H)-one ≥95%

Supplier: Matrix Scientific

047123-500MG 047123-1G
101878-536EA 180 USD
101878-536 101878-538
2-Amino-5-bromopyrimidin-4(3H)-one ≥95%
2-Amino-5-bromopyrimidin-4(3H)-one

Hazard: Irritant

Formula: C₄H₄BrN₃O
Melting Pt: 284…285 °C
CAS Number: 61937-71-1

Order Now

Specification Test Results

Melting point 284-285°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR