4-Fluoro-3-iodobenzaldehyde ≥95%
Supplier: Matrix Scientific
047122-500MG
047122-1G
101878-532EA
747.69
USD
101878-532
101878-534
4-Fluoro-3-iodobenzaldehyde ≥95%
4-Fluoro-3-iodobenzaldehyde
Hazard: Irritant
Formula:
C₇H₄OFI MW: 250.01 g/mol Melting Pt: 67°C |
MDL Number:
MFCD08236850 CAS Number: 227609-88-3 |
Specification Test Results
Melting point | 67°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...