2-(2-Chlorobenzoyl)-3-(dimethylamino)acrylonitrile ≥95%

Supplier: Matrix Scientific

047102-500MG 047102-1G 047102-5G
101878-468EA 278.46 USD
101878-468 101878-470 101878-472
2-(2-Chlorobenzoyl)-3-(dimethylamino)acrylonitrile ≥95%
2-(2-Chlorobenzoyl)-3-(dimethylamino)acrylonitrile

Hazard: Irritant

Formula: C₁₂H₁₁ClN₂O
Melting Pt: 117…119 °C
MDL Number: MFCD09817451
CAS Number: 52200-17-6

Order Now

Specification Test Results

Melting point 117-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR