1-[(2-Nitrophenyl)sulfonyl]piperazine ≥95%

Supplier: Matrix Scientific

047101-500MG 047101-1G
101878-464EA 132.31 USD
101878-464 101878-466
1-[(2-Nitrophenyl)sulfonyl]piperazine ≥95%
1-[(2-Nitrophenyl)sulfonyl]piperazine

Hazard: Irritant

Formula: C₁₀H₁₃N₃O₄S
Melting Pt: 146…148 °C
MDL Number: MFCD00227564
CAS Number: 301331-16-8

Order Now

Specification Test Results

Melting point 146-148°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR