Ethyl-2-chloro-2-[2-(4-methyl-2-nitrophenyl)hydrazono]acetate ≥95%
Supplier: Matrix Scientific
047100-500MG
047100-1G
047100-5G
101878-458EA
181.54
USD
101878-458
101878-460
101878-462
Ethyl-2-chloro-2-[2-(4-methyl-2-nitrophenyl)hydrazono]acetate ≥95%
Ethyl-2-chloro-2-[2-(4-methyl-2-nitrophenyl)hydrazono]acetate
Hazard: Irritant
Formula:
C₁₁H₁₂ClN₃O₄ MW: 285.69 g/mol Melting Pt: 142…143 °C |
MDL Number:
MFCD09864819 CAS Number: 119750-08-2 |
Specification Test Results
Melting point | 142-143°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...