2,4-Dichloro-N'-(2-chloroacetyl)benzenecarbohydrazide ≥95%

Supplier: Matrix Scientific

047098-500MG 047098-1G 047098-5G
101878-448EA 278.46 USD
101878-448 101878-450 101878-452
2,4-Dichloro-N'-(2-chloroacetyl)benzenecarbohydrazide ≥95%
2,4-Dichloro-N'-(2-chloroacetyl)benzenecarbohydrazide

Hazard: Irritant

Formula: C₉H₇Cl₃N₂O₂
Melting Pt: 204…206 °C
MDL Number: MFCD02580953
CAS Number: 184877-74-5

Order Now

Specification Test Results

Melting point 204-206°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR