1-(2-Amino-3-pyridinyl)-1-ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(2-Aminopyridin-3-yl)ethanone, 3-Acetyl-2-aminopyridine

047097-1G 047097-5G
101878-444EA 73.85 USD
101878-444 101878-446
1-(2-Amino-3-pyridinyl)-1-ethanone ≥95%
1-(2-Amino-3-pyridinyl)-1-ethanone

Hazard: Irritant

Formula: C₇H₈N₂O
MW: 136.15 g/mol
Melting Pt: 139…140 °C
MDL Number: MFCD02258895
CAS Number: 65326-33-2

Order Now

Specification Test Results

Melting point 139-140°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR