3-Phenyl-5,6-dihydroimidazo[2,1-b][1,3]thiazole-2-carbaldehyde ≥95%
Supplier: Matrix Scientific
047095-500MG
047095-1G
101878-434EA
747.69
USD
101878-434
101878-436
3-Phenyl-5,6-dihydroimidazo[2,1-b][1,3]thiazole-2-carbaldehyde ≥95%
3-Phenyl-5,6-dihydroimidazo[2,1-b][1,3]thiazole-2-carbaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₀N₂OS MW: 230.29 g/mol Melting Pt: 138…139 °C |
MDL Number:
MFCD09817450 CAS Number: 952182-40-0 |
Specification Test Results
Melting point | 138-139°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...