2-Bromo-3-phenyl-5,6-dihydroimidazo[2,1-b][1,3]thiazole hydrobromide ≥95%
Supplier: Matrix Scientific
047094-500MG
047094-1G
047094-5G
101878-428EA
375.38
USD
101878-428
101878-430
101878-432
2-Bromo-3-phenyl-5,6-dihydroimidazo[2,1-b][1,3]thiazole hydrobromide ≥95%
2-Bromo-3-phenyl-5,6-dihydroimidazo[2,1-b][1,3]thiazole hydrobromide
Hazard: Irritant
Formula:
C₁₁H₁₀Br₂N₂S MW: 362.1 g/mol Melting Pt: 211…222 °C |
MDL Number:
MFCD09817449 CAS Number: 1134611-60-1 |
Specification Test Results
Melting point | 211-222°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...