Methyl-7-bromobenzo[b]thiophene-2-carboxylate ≥95%
Supplier: Matrix Scientific
047092-500MG
047092-1G
101878-418EA
167.69
USD
101878-418
101878-420
Methyl-7-bromobenzo[b]thiophene-2-carboxylate ≥95%
Methyl 7-bromobenzo[b]thiophene-2-carboxylate
Hazard: Irritant
Formula:
C₁₀H₇BrO₂S Melting Pt: 99-102°C |
MDL Number:
MFCD09027095 CAS Number: 550998-53-3 |
Specification Test Results
Melting point | 99-102°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...