Methyl-4-bromobenzo[b]thiophene-2-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-bromobenzo[b]thiophene-2-carboxylate
047091-1G
047091-5G
101878-416EA
92.31
USD
101878-416
102551-362
Methyl-4-bromobenzo[b]thiophene-2-carboxylate ≥95%
Methyl 4-bromobenzo[b]thiophene-2-carboxylate
Hazard: Irritant
Formula:
C₁₀H₇BrO₂S MW: 271.1300 g/mol Melting Pt: 105-108°C |
MDL Number:
MFCD07371547 CAS Number: 360575-29-7 |
Specification Test Results
Melting point | 105-108°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...