Methyl-5-fluorobenzo[b]thiophene-2-carboxylate ≥95%

Supplier: Matrix Scientific

047089-500MG 047089-1G
101878-404EA 278.46 USD
101878-404 101878-406
Methyl-5-fluorobenzo[b]thiophene-2-carboxylate ≥95%
Methyl 5-fluorobenzo[b]thiophene-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₇FO₂S
MW: 210.225 g/mol
Melting Pt: 80…82 °C
MDL Number: MFCD07371535
CAS Number: 154630-32-7

Order Now

Specification Test Results

Melting point 80-82°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR