2-Chloro-1,3-benzothiazole-6-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-6-benzothiazolecarboxylic acid

047088-250MG 047088-1G
101878-400EA 278.46 USD
101878-400 101878-402
2-Chloro-1,3-benzothiazole-6-carboxylic acid ≥95%
2-Chloro-1,3-benzothiazole-6-carboxylic acid

Hazard: Irritant

Formula: C₈H₄ClNO₂S
MW: 213.64 g/mol
Melting Pt: 220…221 °C
MDL Number: MFCD08689736
CAS Number: 3855-95-6

Order Now

Specification Test Results

Melting point 220-221°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR