6-Bromo-2-chlorobenzothiazole ≥95%

Supplier: Matrix Scientific

047086-1G 047086-5G
101878-396EA 15.38 USD
101878-396 101878-398
6-Bromo-2-chlorobenzothiazole ≥95%
6-Bromo-2-chlorobenzothiazole

Hazard: Irritant

Formula: C₇H₃BrClNS
Melting Pt: 93…96 °C
MDL Number: MFCD04971822
CAS Number: 80945-86-4

Order Now

Specification Test Results

Melting point 93-96°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR