2-Hydrazino-6-methylbenzothiazole ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Hydrazinyl-6-methylbenzo[d]thiazole

047085-500MG 047085-1G 047085-5G
101878-390EA 375.38 USD
101878-390 101878-392 101878-394
2-Hydrazino-6-methylbenzothiazole ≥95%
2-Hydrazino-6-methylbenzothiazole

Hazard: Irritant

Formula: C₈H₉N₃S
MW: 179.240 g/mol
Melting Pt: 220°C
MDL Number: MFCD04448800
CAS Number: 20174-69-0

Order Now

Specification Test Results

Melting point 220°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR