4-(2-Oxo-1-imidazolidinyl)benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(2-Oxoimidazolidin-1-yl)benzonitrile

047082-500MG 047082-1G 047082-5G
101878-378EA 375.38 USD
101878-378 101878-380 101878-382
4-(2-Oxo-1-imidazolidinyl)benzenecarbonitrile ≥95%
4-(2-Oxo-1-imidazolidinyl)benzenecarbonitrile

Hazard: Irritant

Formula: C₁₀H₉N₃O
MW: 187.2 g/mol
Melting Pt: 179…181 °C
MDL Number: MFCD08689735
CAS Number: 144655-81-2

Order Now

Specification Test Results

Melting point 179-181°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR