Methyl-5,6-dihydro-4H-cyclopenta[b]thiophene-2-carboxylate ≥95%

Supplier: Matrix Scientific

047080-500MG 047080-1G 047080-5G
101878-372EA 181.54 USD
101878-372 101878-374 101878-376
Methyl-5,6-dihydro-4H-cyclopenta[b]thiophene-2-carboxylate ≥95%
Formula: C₉H₁₀O₂S
Melting Pt: 60…62 °C
MDL Number: MFCD01927210
CAS Number: 353269-02-0

Order Now

Specification Test Results

Melting point 60-62°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR