1-(3,4-Dichlorophenyl)tetrahydro-2H-imidazol-2-one ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(3,4-Dichlorophenyl)imidazolidin-2-one

047079-500MG 047079-1G 047079-5G
101878-366EA 375.38 USD
101878-366 101878-368 101878-370
1-(3,4-Dichlorophenyl)tetrahydro-2H-imidazol-2-one ≥95%
1-(3,4-Dichlorophenyl)tetrahydro-2H-imidazol-2-one

Hazard: Irritant

Formula: C₉H₈Cl₂N₂O
MW: 231.08 g/mol
Melting Pt: 185…187 °C
MDL Number: MFCD08689734
CAS Number: 52420-29-8

Order Now

Specification Test Results

Melting point 185-187°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR