N-(2-Chloroethyl)-N'-[2-(2-pyridinyl)ethyl]urea ≥95%
Supplier: Matrix Scientific
047078-500MG
047078-1G
047078-5G
101878-360EA
278.46
USD
101878-360
101878-362
101878-364
N-(2-Chloroethyl)-N'-[2-(2-pyridinyl)ethyl]urea ≥95%
N-(2-Chloroethyl)-N'-[2-(2-pyridinyl)ethyl]urea
Hazard: Irritant
Formula:
C₁₀H₁₄ClN₃O MW: 227.7 g/mol Melting Pt: 112…114 °C |
MDL Number:
MFCD08689733 CAS Number: 42471-42-1 |
Specification Test Results
Melting point | 112-114°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...