8-Benzyl-1,2,8-triazaspiro[4.5]decan-3-one ≥95%

Supplier: Matrix Scientific

047076-500MG 047076-1G 047076-5G
101878-348EA 375.38 USD
101878-348 101878-350 101878-352
8-Benzyl-1,2,8-triazaspiro[4.5]decan-3-one ≥95%
8-Benzyl-1,2,8-triazaspiro[4.5]decan-3-one

Hazard: Irritant

Formula: C₁₄H₁₉N₃O
Melting Pt: 187 °C
MDL Number: MFCD08689731
CAS Number: 785782-25-4

Order Now

Specification Test Results

Melting point 187°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR