N-(2-Chloroethyl)-N'-(3,4-dichlorophenyl)urea ≥95%

Supplier: Matrix Scientific

047074-500MG 047074-1G 047074-5G
101878-336EA 278.46 USD
101878-336 101878-338 101878-340
N-(2-Chloroethyl)-N'-(3,4-dichlorophenyl)urea ≥95%
N-(2-Chloroethyl)-N'-(3,4-dichlorophenyl)urea

Hazard: Irritant

Formula: C₉H₉Cl₃N₂O
MW: 267.54 g/mol
Melting Pt: 120…122 °C
MDL Number: MFCD01676684
CAS Number: 15145-37-6

Order Now

Specification Test Results

Melting point 120-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR