N-(2-Chloroethyl)-N'-(3-pyridinyl)urea ≥95%
Supplier: Matrix Scientific
047071-500MG
047071-1G
047071-5G
101878-320EA
278.46
USD
101878-320
101878-322
101878-324
N-(2-Chloroethyl)-N'-(3-pyridinyl)urea ≥95%
N-(2-Chloroethyl)-N'-(3-pyridinyl)urea
Hazard: Irritant
Formula:
C₈H₁₀ClN₃O MW: 199.64 g/mol Melting Pt: 143…144 °C |
MDL Number:
MFCD08689730 CAS Number: 13908-58-2 |
Specification Test Results
Melting point | 143-144°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...