N-(2-Chloroethyl)-N'-(2,4-dimethylphenyl)urea ≥95%

Supplier: Matrix Scientific

047069-500MG 047069-1G 047069-5G
101878-308EA 278.46 USD
101878-308 101878-310 101878-312
N-(2-Chloroethyl)-N'-(2,4-dimethylphenyl)urea ≥95%
N-(2-Chloroethyl)-N'-(2,4-dimethylphenyl)urea

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O
Melting Pt: 169…170 °C
MDL Number: MFCD00444204
CAS Number: 162376-66-1

Order Now

Specification Test Results

Melting point 169-170°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR