N-(2-Chloroethyl)-N'-[4-(methylsulfanyl)phenyl]urea ≥95%
Supplier: Matrix Scientific
Synonyms:
N-(2-Chloroethyl)-N'-[4-(methylthio)phenyl]urea
047067-500MG
047067-1G
047067-5G
101878-302EA
278.46
USD
101878-302
101878-304
101878-306
N-(2-Chloroethyl)-N'-[4-(methylsulfanyl)phenyl]urea ≥95%
N-(2-Chloroethyl)-N'-[4-(methylsulfanyl)phenyl]urea
Hazard: Irritant
Formula:
C₁₀H₁₃ClN₂OS MW: 244.74 g/mol Melting Pt: 130…131 °C |
MDL Number:
MFCD08689728 CAS Number: 13908-50-4 |
Specification Test Results
Melting point | 130-131°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...