N-(2-Chloroethyl)-N'-(3-chloro-4-methylphenyl)urea ≥95%

Supplier: Matrix Scientific

047066-500MG 047066-1G 047066-5G
101878-296EA 181.54 USD
101878-296 101878-298 101878-300
N-(2-Chloroethyl)-N'-(3-chloro-4-methylphenyl)urea ≥95%
N-(2-Chloroethyl)-N'-(3-chloro-4-methylphenyl)urea

Hazard: Irritant

Formula: C₁₀H₁₂Cl₂N₂O
Melting Pt: 147…148 °C
MDL Number: MFCD08689727
CAS Number: 937601-77-9

Order Now

Specification Test Results

Melting point 147-148°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR