1-(4-Methoxyphenyl)tetrahydro-2H-imidazol-2-one ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(4-Methoxyphenyl)imidazolidin-2-one

047064-500MG 047064-1G 047064-5G
101878-286EA 160 USD
101878-286 101878-288 101878-290
1-(4-Methoxyphenyl)tetrahydro-2H-imidazol-2-one ≥95%
1-(4-Methoxyphenyl)tetrahydro-2H-imidazol-2-one

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₂
MW: 192.22 g/mol
Melting Pt: 210…212 °C
MDL Number: MFCD01720969
CAS Number: 62868-39-7

Order Now

Specification Test Results

Melting point 210-212°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR