tert-Butyl-4-{2-[4-(hydrazinocarbonyl)piperidino]ethyl}tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-{2-[4-(Hydrazinocarbonyl)piperidin-1-yl]ethyl}piperazine, N1-BOC protected

047063-500MG 047063-1G 047063-5G
101878-280EA 556.92 USD
101878-280 101878-282 101878-284
tert-Butyl-4-{2-[4-(hydrazinocarbonyl)piperidino]ethyl}tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-{2-[4-(hydrazinocarbonyl)piperidino]ethyl}tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₁₇H₃₃N₅O₃
MDL Number: MFCD06808601
CAS Number: 874831-75-1

Order Now

Specification Test Results

Melting point 136-138°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR