Methyl-4-amino-3-hydroxybenzoate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-amino-3-hydroxybenzoate
047061-5G
047061-25G
101878-278EA
56.92
USD
101878-278
102551-360
Methyl-4-amino-3-hydroxybenzoate ≥95%
Methyl 4-amino-3-hydroxybenzoate
Hazard: Irritant
Formula:
C₈H₉NO₃ MW: 167.17 g/mol Melting Pt: 118-120°C |
MDL Number:
MFCD00017093 CAS Number: 63435-16-5 |
Specification Test Results
Melting point | 118-120°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...