2-(Chloromethyl)-1-(4-chlorophenoxy)-4-nitrobenzene ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(4-Chlorophenoxy)-5-nitrobenzyl chloride
047057-500MG
047057-1G
047057-5G
101878-256EA
375.38
USD
101878-256
101878-258
101878-260
2-(Chloromethyl)-1-(4-chlorophenoxy)-4-nitrobenzene ≥95%
2-(Chloromethyl)-1-(4-chlorophenoxy)-4-nitrobenzene
Hazard: Irritant
Formula:
C₁₃H₉Cl₂NO₃ MW: 298.13 g/mol Melting Pt: 90…92 °C |
MDL Number:
MFCD08689723 CAS Number: 924862-19-1 |
Specification Test Results
Melting point | 90-92°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...