Methyl-2-(chloromethyl)-1,3-benzoxazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

047056-500MG 047056-1G
101878-252EA 346.15 USD
101878-252 101878-254
Methyl-2-(chloromethyl)-1,3-benzoxazole-5-carboxylate ≥95%
Methyl-2-(chloromethyl)-1,3-benzoxazole-5-carboxylate

Hazard: Irritant

Formula: C₁₀H₈ClNO₃
MW: 225.63 g/mol
Melting Pt: 90…91 °C
MDL Number: MFCD08689722
CAS Number: 924862-18-0

Order Now

Specification Test Results

Melting point 90-91°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR