tert-Butyl-2,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 6,7-dihydro-2H-pyrazolo[4,3-c]pyridine-5(4H)-carboxylate, 2,4,6,7-Tetrahydro-5H-pyrazolo[4,3-c]pyridine, N5-BOC protected

047054-250MG 047054-1G 047054-5G
101878-240EA 121.54 USD
101878-240 101878-242 101878-244
tert-Butyl-2,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate ≥95%
tert-Butyl-2,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₇N₃O₂
MW: 223.28 g/mol
Melting Pt: 127…128 °C
MDL Number: MFCD08689720
CAS Number: 924869-27-2

Order Now

Specification Test Results

Melting point 127-128°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR